Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-3-fluorobenzene 98%

Supplier: AMBEED, INC
Danger

A258216-10G A258216-25G
77113-606EA 5.15 USD InStock
77113-606 77113-608
1-Bromo-3-fluorobenzene 98%
1-Bromo-3-fluorobenzene
Precaución: for Research Use Only


Formula: C₆H₄BrF
MW: 175 g/mol
Boiling Pt: 148
Melting Pt: -8
Flash Pt: 46
MDL Number: MFCD00000326
CAS Number: 1073-06-9
UN: 1993
ADR: 3 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty