Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Foscarnet sodium 97%

Supplier: AMBEED, INC
Warning

A262254-250MG A262254-5G A262254-25G
77113-708EA 6.18 USD InStock
77113-708 77612-512 77685-747
Foscarnet sodium 97%
Foscarnet sodium
Precaución: for Research Use Only


Formula: CNa₃O₅P
MW: 191.95 g/mol
MDL Number: MFCD00013973
CAS Number: 63585-09-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty