Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Chloro-2,3-dihydro-1H-indole hydrochloride 95%

Supplier: AMBEED, INC
Warning

A264546-10G A264546-1G A264546-250MG A264546-5G
77113-766EA 328.57 USD InStock
77113-766 77113-768 77113-770 77113-772
6-Chloro-2,3-dihydro-1H-indole hydrochloride 95%
6-Chloro-2,3-dihydro-1H-indole hydrochloride
Precaución: for Research Use Only


Formula: C₈H₉Cl₂N
MDL Number: MFCD09026834
CAS Number: 89978-84-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty