Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzylacetone 98%

Supplier: AMBEED, INC
Warning

A265752-1000G A265752-100G A265752-25G A265752-500G
77113-796EA 81.37 USD InStock
77113-796 77113-798 77113-800 77113-802
Benzylacetone 98%
Benzylacetone
Precaución: for Research Use Only


Formula: (C₆H₅)CH₂CH₂C(O)CH₃
MDL Number: MFCD00008790
CAS Number: 2550-26-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty