Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(4-Ethylphenyl)-3-phenylurea 98%

Supplier: AMBEED, INC
Danger

A267572-10MG A267572-1MG A267572-25MG A267572-5MG
77113-838EA 36.05 USD InStock
77113-838 77113-840 77113-842 77113-844
1-(4-Ethylphenyl)-3-phenylurea 98%
1-(4-Ethylphenyl)-3-phenylurea
Precaución: for Research Use Only


Formula: C₁₅H₁₆N₂O
MDL Number: MFCD00158246
CAS Number: 200134-22-1
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty