Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

DL-Propargylglycine 97%

Supplier: AMBEED, INC
Warning

A268199-1G A268199-250MG A268199-5G
77113-854EA 36.05 USD InStock
77113-854 77113-856 77113-858
DL-Propargylglycine 97%
DL-Propargylglycine
Precaución: for Research Use Only


Formula: C₅H₇NO₂
MDL Number: MFCD00056728
CAS Number: 64165-64-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty