Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl cyanoacetate 95%

Supplier: AMBEED, INC
Warning

A270018-1000G A270018-100G A270018-25G A270018-500G
77113-882EA 27.81 USD InStock
77113-882 77113-884 77113-886 77113-888
Methyl cyanoacetate 95%
Methyl cyanoacetate
Precaución: for Research Use Only


Formula: C₄H₅NO₂
MDL Number: MFCD00001939
CAS Number: 105-34-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty