Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dimethoxyaniline 95%

Supplier: AMBEED, INC
Warning

A277241-25G A277241-500G A277241-5G
77113-980EA 25.76 USD InStock
77113-980 77113-982 77113-984
3,5-Dimethoxyaniline 95%
3,5-Dimethoxyaniline
Precaución: for Research Use Only


Formula: C₈H₁₁NO₂
MW: 153.18 g/mol
Boiling Pt: 303
MDL Number: MFCD00008392
CAS Number: 10272-07-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty