Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-D-Lys(Fmoc)-OH 96%

Supplier: AMBEED, INC
Warning

A279759-250MG A279759-25G A279759-5G A279759-100G A279759-1G
77114-040EA 11.33 USD InStock
77114-040 77114-042 77114-044 77685-787 77685-788
Fmoc-D-Lys(Fmoc)-OH 96%
Fmoc-D-Lys(Fmoc)-OH
Precaución: for Research Use Only


Formula: C₃₆H₃₄N₂O₆
MW: 590.676 g/mol
MDL Number: MFCD00270743
CAS Number: 75932-02-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty