Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-2,6-difluorobenzene 98%

Supplier: AMBEED, INC
Danger

A280971-10G A280971-1G A280971-25G A280971-5G A280971-100G
77114-058EA 8.23 USD InStock
77114-058 77114-060 77114-062 77114-064 77685-794
1-Bromo-2,6-difluorobenzene 98%
1-Bromo-2,6-difluorobenzene
Precaución: for Research Use Only


Formula: C₆H₃BrF₂
MDL Number: MFCD00009894
CAS Number: 64248-56-2
UN: 1993
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty