Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methyl-1H-pyrrolo[2,3-b]pyridine 98%

Supplier: AMBEED, INC
Warning

A281165-10G A281165-1G A281165-250MG A281165-25G A281165-5G
77114-070EA 118.45 USD InStock
77114-070 77114-072 77114-074 77114-076 77114-078
2-Methyl-1H-pyrrolo[2,3-b]pyridine 98%
2-Methyl-1H-pyrrolo[2,3-b]pyridine
Precaución: for Research Use Only


Formula: C₈H₈N₂
MDL Number: MFCD08669524
CAS Number: 23612-48-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty