Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Lansoprazole 98%

Supplier: AMBEED, INC
Warning

A281446-100G A281446-100MG A281446-1G A281446-25G A281446-5G A281446-1MG
77114-080EA 323.42 USD InStock
77114-080 77114-082 77114-084 77114-086 77114-088 77685-795
Lansoprazole 98%
Lansoprazole
Precaución: for Research Use Only


Formula: C₁₆H₁₄F₃N₃O₂S
MDL Number: MFCD00866873
CAS Number: 103577-45-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty