Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Bis(trifluoromethyl)anisole 97%

Supplier: AMBEED, INC
Danger

A281879-10G A281879-25G A281879-5G
77114-092EA 36.05 USD InStock
77114-092 77114-094 77114-096
3,5-Bis(trifluoromethyl)anisole 97%
3,5-Bis(trifluoromethyl)anisole
Precaución: for Research Use Only


Formula: C₉H₆F₆O
MDL Number: MFCD00728736
CAS Number: 349-60-0
UN: 1993
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty