Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4,5-Trifluoro-4-hydroxybenzoic acid 98%

Supplier: AMBEED, INC
Warning

A286379-1G A286379-25G A286379-5G
77114-158EA 10.31 USD InStock
77114-158 77114-160 77114-162
2,4,5-Trifluoro-4-hydroxybenzoic acid 98%
2,4,5-Trifluoro-4-hydroxybenzoic acid
Precaución: for Research Use Only


Formula: C₇H₃F₃O₃
MDL Number: MFCD00800378
CAS Number: 116751-24-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty