Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

MI-2, MALT1 Inhibitor 98+%

Supplier: AMBEED, INC
Warning

A286737-100MG A286737-10MG A286737-1MG A286737-50MG A286737-5MG
77114-168EA 115.36 USD InStock
77114-168 77114-170 77114-172 77114-174 77114-176
MI-2, MALT1 Inhibitor 98+%
MI-2, MALT1 Inhibitor
Precaución: for Research Use Only


Formula: C₁₉H₁₇Cl₃N₄O₃
MDL Number: MFCD19754074
CAS Number: 1047953-91-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty