Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

m,α,α,α-Tetrafluoroanisole 98%

Supplier: AMBEED, INC
Warning

A287308-100G A287308-1G A287308-25G A287308-5G
77114-186EA 200.85 USD InStock
77114-186 77114-188 77114-190 77114-192
m,α,α,α-Tetrafluoroanisole 98%
m,α,α,α-Tetrafluoroanisole
Precaución: for Research Use Only


Formula: C₇H₄F₄O
MDL Number: MFCD00236323
CAS Number: 1077-01-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty