Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-3,4-difluoro-1-nitrobenzene 98%

Supplier: AMBEED, INC
Warning

A296039-100G A296039-10G A296039-1G A296039-25G A296039-5G
77114-374EA 200.85 USD InStock
77114-374 77114-376 77114-378 77114-380 77114-382
2-Bromo-3,4-difluoro-1-nitrobenzene 98%
2-Bromo-3,4-difluoro-1-nitrobenzene
Precaución: for Research Use Only


Formula: C₆H₂BrF₂NO₂
MDL Number: MFCD01820590
CAS Number: 350699-92-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty