Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-5-methoxybenzyl bromide 98%

Supplier: AMBEED, INC
Danger

A296738-100MG A296738-1G A296738-250MG A296738-5G
77114-398EA 19.57 USD InStock
77114-398 77114-400 77114-402 77114-404
3-Fluoro-5-methoxybenzyl bromide 98%
3-Fluoro-5-methoxybenzyl bromide
Precaución: for Research Use Only


Formula: C₈H₈BrFO
MDL Number: MFCD08741378
CAS Number: 914637-29-9
UN: 3261
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty