Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Aminomorpholine 95%

Supplier: AMBEED, INC
Danger

A306582-10G A306582-1G A306582-25G A306582-5G
77114-544EA 22.66 USD InStock
77114-544 77114-546 77114-548 77114-550
4-Aminomorpholine 95%
4-Aminomorpholine
Precaución: for Research Use Only


Formula: C₄H₁₀N₂O
MW: 102.14 g/mol
MDL Number: MFCD00006174
CAS Number: 4319-49-7
UN: 2733
ADR: 3,8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty