Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl-4-[(2E)-3-(dimethylamino)prop-2-enoyl]piperidine-1-carboxylate 95%

Supplier: AMBEED, INC
Danger

A309897-1G A309897-250MG
77114-588EA 121.54 USD InStock
77114-588 77114-590
tert-Butyl-4-[(2E)-3-(dimethylamino)prop-2-enoyl]piperidine-1-carboxylate 95%
tert-Butyl-4-[(2E)-3-(dimethylamino)prop-2-enoyl]piperidine-1-carboxylate
Precaución: for Research Use Only


Formula: C₁₅H₂₆N₂O₃
MDL Number: MFCD08898850
CAS Number: 960201-86-9
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty