Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Acetylthiophene 98%

Supplier: AMBEED, INC
Warning

A310778-100G A310778-25G A310778-500G
77114-612EA 30.92 USD InStock
77114-612 77114-614 77114-616
2-Acetylthiophene 98%
2-Acetylthiophene
Precaución: for Research Use Only


Formula: C₆H₆OS
MW: 126.18 g/mol
Boiling Pt: 214
Melting Pt: 9
MDL Number: MFCD00005442
CAS Number: 88-15-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty