Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Diaminepyrimidine 95%

Supplier: AMBEED, INC
Warning

A323490-100MG A323490-1G A323490-250MG A323490-5G
77114-746EA 37.08 USD InStock
77114-746 77114-748 77114-750 77114-752
2,5-Diaminepyrimidine 95%
2,5-Diaminepyrimidine
Precaución: for Research Use Only


Formula: C₄H₆N₄
MDL Number: MFCD00129726
CAS Number: 22715-27-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty