Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Oxabicyclo[3.2.0]heptane-2,4-dione 95%

Supplier: AMBEED, INC
Warning

A328079-10G A328079-1G A328079-250MG A328079-25G A328079-5G
77114-840EA 73.14 USD InStock
77114-840 77114-842 77114-844 77114-846 77593-940
3-Oxabicyclo[3.2.0]heptane-2,4-dione 95%
3-Oxabicyclo[3.2.0]heptane-2,4-dione
Precaución: for Research Use Only


Formula: C₆H₆O₃
MW: 126.11 g/mol
MDL Number: MFCD00799396
CAS Number: 4462-96-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty