Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Nitro-2,4,6-triaminopyrimidine 95%

Supplier: AMBEED, INC
Warning

A331330-100MG A331330-1G A331330-250MG
77114-878EA 77.25 USD InStock
77114-878 77114-880 77114-882
5-Nitro-2,4,6-triaminopyrimidine 95%
5-Nitro-2,4,6-triaminopyrimidine
Precaución: for Research Use Only


Formula: C₄H₆N₆O₂
MDL Number: MFCD00047376
CAS Number: 24867-36-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty