Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(Benzyloxy)propan-2-ol 97%

Supplier: AMBEED, INC
Warning

A337427-10G A337427-1G A337427-250MG A337427-25G A337427-5G
77114-982EA 148.32 USD InStock
77114-982 77114-984 77114-986 77114-988 77114-990
1-(Benzyloxy)propan-2-ol 97%
1-(Benzyloxy)propan-2-ol
Precaución: for Research Use Only


Formula: C₁₀H₁₄O₂
MW: 166.21 g/mol
MDL Number: MFCD00085681
CAS Number: 13807-91-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty