Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-2-fluoro-4-iodopyridine 97%

Supplier: AMBEED, INC
Warning

A343513-10G A343513-1G A343513-250MG A343513-5G A343513-25G
77115-050EA 77.24 USD InStock
77115-050 77115-052 77115-054 77685-982 77685-981
5-Chloro-2-fluoro-4-iodopyridine 97%
5-Chloro-2-fluoro-4-iodopyridine
Precaución: for Research Use Only


Formula: C₅H₂ClFIN
MDL Number: MFCD04112566
CAS Number: 659731-48-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty