Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-(6-Methoxyquinolin-4-yl)((1S,2S,4S,5R)-5-vinylquinuclidin-2-yl)methyl acetate 97% stabilized

Supplier: AMBEED, INC
Warning

Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A348403-100MG A348403-1G A348403-250MG
77115-092EA 30.9 USD InStock
77115-092 77115-094 77115-096
(R)-(6-Methoxyquinolin-4-yl)((1S,2S,4S,5R)-5-vinylquinuclidin-2-yl)methyl acetate 97% stabilized
(R)-(6-Methoxyquinolin-4-yl)((1S,2S,4S,5R)-5-vinylquinuclidin-2-yl)methyl acetate
Precaución: for Research Use Only


Formula: C₂₂H₂₆N₂O₃
MDL Number: MFCD00050910
CAS Number: 18797-86-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty