Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-(trifluoromethoxy)benzenesulfonylchloride 98%

Supplier: AMBEED, INC
Danger

A351461-1G A351461-25G A351461-5G
77115-120EA 14.42 USD InStock
77115-120 77115-122 77115-124
4-Bromo-2-(trifluoromethoxy)benzenesulfonylchloride 98%
4-Bromo-2-(trifluoromethoxy)benzenesulfonylchloride
Precaución: for Research Use Only


Formula: C₇H₃BrClF₃O₃S
MW: 339.52 g/mol
MDL Number: MFCD00179336
CAS Number: 175278-14-5
UN: 3265
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty