Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(Diethoxyphosphoryl)methyl-4-methylbenzenesulfonate 98%

Supplier: AMBEED, INC
Warning

A351651-1000G A351651-100G A351651-10G A351651-25G A351651-500G A351651-5G
77115-126EA 78.28 USD InStock
77115-126 77115-128 77115-130 77115-132 77115-134 77115-136
(Diethoxyphosphoryl)methyl-4-methylbenzenesulfonate 98%
(Diethoxyphosphoryl)methyl-4-methylbenzenesulfonate
Precaución: for Research Use Only


Formula: C₁₂H₁₉O₆PS
MW: 322.3100 g/mol
MDL Number: MFCD03412078
CAS Number: 31618-90-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty