Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-fluorophenylacetic acid 98%

Supplier: AMBEED, INC
Warning

A354116-100G A354116-10G A354116-1G A354116-25G A354116-500G A354116-5G
77115-148EA 133.91 USD InStock
77115-148 77115-150 77115-152 77115-154 77115-156 77115-158
4-Bromo-2-fluorophenylacetic acid 98%
4-Bromo-2-fluorophenylacetic acid
Precaución: for Research Use Only


Formula: C₈H₆BrFO₂
MW: 233.03 g/mol
MDL Number: MFCD09032952
CAS Number: 114897-92-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty