Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dasatinib 98%

Supplier: AMBEED, INC
Danger

A355193-100MG A355193-1G A355193-250MG A355193-5G A355193-100G
77115-176EA 8.24 USD InStock
77115-176 77115-178 77115-180 77115-182 77686-011
Dasatinib 98%
Dasatinib
Precaución: for Research Use Only


Formula: C₂₂H₂₆ClN₇O₂S
MW: 488 g/mol
MDL Number: MFCD11046566
CAS Number: 302962-49-8
UN: 3077
ADR: 9 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty