Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Disodium-2,6-naphthalenedisulfonate 97%

Supplier: AMBEED, INC
Warning

A355470-1000G A355470-100G A355470-25G A355470-500G
77115-184EA 72.1 USD InStock
77115-184 77115-186 77115-188 77115-190
Disodium-2,6-naphthalenedisulfonate 97%
Disodium-2,6-naphthalenedisulfonate
Precaución: for Research Use Only


Formula: C₁₀H₈O₆S₂
MW: 332.25 g/mol
MDL Number: MFCD00004092
CAS Number: 1655-45-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty