Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methoxy-2-nitropyridine 98%

Supplier: AMBEED, INC
Warning

A361015-100G A361015-10G A361015-25G A361015-500G A361015-5G
77115-298EA 21.63 USD InStock
77115-298 77115-300 77115-302 77115-304 77115-306
3-Methoxy-2-nitropyridine 98%
3-Methoxy-2-nitropyridine
Precaución: for Research Use Only


Formula: C₆H₆N₂O₃
MDL Number: MFCD00222276
CAS Number: 20265-37-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty