Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(4-Chlorophenyl)hexan-1-one 95%

Supplier: AMBEED, INC
Danger

A378117-100MG A378117-1G A378117-250MG
77115-468EA 11.33 USD InStock
77115-468 77115-470 77115-472
1-(4-Chlorophenyl)hexan-1-one 95%
1-(4-Chlorophenyl)hexan-1-one
Precaución: for Research Use Only


Formula: C₁₂H₁₅ClO
MDL Number: MFCD00090186
CAS Number: 7295-50-3
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty