Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium 4-(trifluoromethoxy) phenyltrifluoroborate 95+%

Supplier: AMBEED, INC
Warning

A383380-1G A383380-250MG A383380-5G
77115-528EA 22.66 USD InStock
77115-528 77115-530 77115-532
Potassium 4-(trifluoromethoxy) phenyltrifluoroborate 95+%
Potassium 4-(trifluoromethoxy) phenyltrifluoroborate
Precaución: for Research Use Only


Formula: C₇H₄BF₆O·K
MW: 268.006 g/mol
MDL Number: MFCD18861588
CAS Number: 1394827-04-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty