Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

trans-4-Methylcyclohexanamine hydrochloride 97%

Supplier: AMBEED, INC
Warning

A392261-10G A392261-25G A392261-5G
77115-652EA 6.18 USD InStock
77115-652 77115-654 77115-656
trans-4-Methylcyclohexanamine hydrochloride 97%
trans-4-Methylcyclohexanamine hydrochloride
Precaución: for Research Use Only


Formula: C₇H₁₆ClN
MDL Number: MFCD07368256
CAS Number: 33483-65-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty