Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

α,3-Dibromo-4-fluorotoluene 98%

Supplier: AMBEED, INC
Danger

A402346-10G A402346-25G A402346-5G
77115-738EA 35.02 USD InStock
77115-738 77115-740 77115-742
α,3-Dibromo-4-fluorotoluene 98%
α,3-Dibromo-4-fluorotoluene
Precaución: for Research Use Only


Formula: C₇H₅Br₂F
MW: 267.92 g/mol
MDL Number: MFCD09037367
CAS Number: 78239-71-1
UN: 3261
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty