Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,N'-Diphenylformamidine 98%

Supplier: AMBEED, INC
Warning

A409052-100G A409052-25G A409052-500G A409052-5G
77115-804EA 86.52 USD InStock
77115-804 77115-806 77115-808 77115-810
N,N'-Diphenylformamidine 98%
N,N'-Diphenylformamidine


Formula: C₁₃H₁₂N₂
MW: 196.25 g/mol
MDL Number: MFCD00003026
CAS Number: 622-15-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty