Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-(+)-2-Amino-1,1-diphenyl-1-propanol 98%

Supplier: AMBEED, INC
Warning

A411771-10G A411771-5G A411771-250MG
77115-832EA 211.15 USD InStock
77115-832 77115-834 77686-136
(R)-(+)-2-Amino-1,1-diphenyl-1-propanol 98%
(R)-(+)-2-Amino-1,1-diphenyl-1-propanol
Precaución: for Research Use Only


Formula: C₁₅H₁₇NO
MDL Number: MFCD02677967
CAS Number: 78603-93-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty