Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

OSI-930 98+%

Supplier: AMBEED, INC
Warning

A412992-10MG A412992-1MG A412992-2MG A412992-50MG A412992-5MG
77115-848EA 58.71 USD InStock
77115-848 77115-850 77115-852 77115-854 77115-856
OSI-930 98+%
OSI-930
Precaución: for Research Use Only


Formula: C₂₂H₁₆F₃N₃O₂S
MW: 443.44 g/mol
MDL Number: MFCD11100358
CAS Number: 728033-96-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty