Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2,2-difluoro-1,3-benzodioxole 98%

Supplier: AMBEED, INC
Warning

A421523-1G A421523-250MG A421523-25G A421523-5G
77115-936EA 15.44 USD InStock
77115-936 77115-938 77115-940 77115-942
4-Bromo-2,2-difluoro-1,3-benzodioxole 98%
4-Bromo-2,2-difluoro-1,3-benzodioxole
Precaución: for Research Use Only


Formula: C₇H₃BrF₂O₂
MDL Number: MFCD01631385
CAS Number: 144584-66-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty