Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

CVT-313 98%

Supplier: AMBEED, INC
Warning

A421689-10MG A421689-1MG A421689-25MG A421689-50MG A421689-5MG
77115-944EA 73.14 USD InStock
77115-944 77115-946 77115-948 77115-950 77115-952
CVT-313 98%
CVT-313
Precaución: for Research Use Only


Formula: C₂₀H₂₈N₆O₃
MW: 400.4747 g/mol
MDL Number: MFCD06411412
CAS Number: 199986-75-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty