Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dimethyl-8-hydroxyquinoline 97%

Supplier: AMBEED, INC
Danger

A445169-100MG A445169-250MG A445169-5G A445169-1G
77116-166EA 48.41 USD InStock
77116-166 77116-168 77116-170 77686-207
2,4-Dimethyl-8-hydroxyquinoline 97%
2,4-Dimethyl-8-hydroxyquinoline
Precaución: for Research Use Only


Formula: C₁₁H₁₁NO
MDL Number: MFCD09743812
CAS Number: 115310-98-0
UN: 1759
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty