Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3,4-Trifluorobenzenesulfonyl chloride 95%

Supplier: AMBEED, INC
Danger

A466251-10G A466251-1G
77116-332EA 23.69 USD InStock
77116-332 77116-334
2,3,4-Trifluorobenzenesulfonyl chloride 95%
Trifluorobenzenesulphonyl chloride
Precaución: for Research Use Only


Formula: C₆H₂ClF₃O₂S
MDL Number: MFCD00102573
CAS Number: 175278-08-7
UN: 3265
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty