Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,2,4-Triaminobenzene dihydrochloride 95%

Supplier: AMBEED, INC
Warning

A482416-100G A482416-1G A482416-25G
77116-426EA 405.81 USD InStock
77116-426 77116-428 77116-430
1,2,4-Triaminobenzene dihydrochloride 95%
1,2,4-Triaminobenzene dihydrochloride
Precaución: for Research Use Only


Formula: C₆H₉N₃
MDL Number: MFCD00016619
CAS Number: 615-47-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty