Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Dibromohexanediamide 95%

Supplier: AMBEED, INC
Warning

A488228-100MG A488228-250MG
77116-442EA 17.52 USD InStock
77116-442 77116-444
2,5-Dibromohexanediamide 95%
2,5-Dibromohexanediamide
Precaución: for Research Use Only


Formula: C₆H₁₀Br₂N₂O₂
MW: 301.96 g/mol
MDL Number: MFCD27976335
CAS Number: 99584-96-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty