Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Chloromethyl)benzoyl chloride 98%

Supplier: AMBEED, INC
Danger

A511182-1G A511182-25G A511182-5G
77116-628EA 8.24 USD InStock
77116-628 77116-630 77116-632
3-(Chloromethyl)benzoyl chloride 98%
3-(Chloromethyl)benzoyl chloride
Precaución: for Research Use Only


Formula: C₈H₆Cl₂O
MW: 189.04
Boiling Pt: 268
Melting Pt: 4
Density: 1.331
MDL Number: MFCD00000682
CAS Number: 63024-77-1
UN: 3265
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty