Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N6-Cyclohexyl-N2-(2-methyl-4-morpholinophenyl)-9H-purine-2,6-diamine 98%

Supplier: AMBEED, INC
Warning

A519540-10MG A519540-1MG A519540-2MG A519540-50MG A519540-5MG
77116-670EA 35.02 USD InStock
77116-670 77116-672 77116-674 77116-676 77116-678
N6-Cyclohexyl-N2-(2-methyl-4-morpholinophenyl)-9H-purine-2,6-diamine 98%
N6-Cyclohexyl-N2-(2-methyl-4-morpholinophenyl)-9H-purine-2,6-diamine
Precaución: for Research Use Only


Formula: C₂₂H₂₉N₇O
MDL Number: MFCD28099811
CAS Number: 1246529-32-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty