Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Difluoroanisole 98%

Supplier: AMBEED, INC
Danger

A530867-25G A530867-5G
77116-734EA 30.9 USD InStock
77116-734 77116-736
3,4-Difluoroanisole 98%
3,4-Difluoroanisole
Precaución: for Research Use Only


Formula: C₇H₆F₂O
MW: 144.12
Boiling Pt: 139
Melting Pt: -30
Density: 1.23
MDL Number: MFCD00010730
CAS Number: 115144-40-6
UN: 1993
ADR: 3 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty