Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium 1-benzofuran-2-yltrifluoroboranuide 98%

Supplier: AMBEED, INC
Warning

A534537-1G A534537-25G A534537-5G
77116-760EA 37.07 USD InStock
77116-760 77116-762 77116-764
Potassium 1-benzofuran-2-yltrifluoroboranuide 98%
Potassium 1-benzofuran-2-yltrifluoroboranuide
Precaución: for Research Use Only


Formula: C₈H₅BF₃O·K
MDL Number: MFCD11052671
CAS Number: 929626-27-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty